The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027870.3 | NP_082146.2 | 1 - 1 | 53 | 104 |
GenBank mRNA | CJ071609.1 | - | 1 - 1 | 54 | 104 |