The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011931.3 | NP_036061.1 | 20 - 20 | 4345 | 100 |
GenBank mRNA | AK039275.1 | - | 1 - 1 | 280 | 100 |
AK042587.1 | - | 1 - 1 | 1263 | 100 | |
BC082804.1 | - | 20 - 20 | 4345 | 100 |