Assay Design
The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Gene Transcripts
- Gene Symbol
- Gm15299
- Entrez Gene ID
-
100041811
- Gene Name
- predicted pseudogene 15299
- Gene Aliases
-
ENSMUSG00000074438, OTTMUSG00000019793
- Chromosome Location
-
Chr.8: 21315546 - 21316387 on Build GRCm38
- Gene Symbol
- Defa25
- Entrez Gene ID
-
13236
- Gene Name
- defensin, alpha, 25
- Gene Aliases
-
Defcr-2, Defcr2, Defcr25
- Chromosome Location
-
Chr.8: 21084442 - 21085285 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_007849.1 |
NP_031875.1 |
1 - 1 |
153 |
94 |
GenBank mRNA |
AY746430.1 |
- |
1 - 1 |
153 |
94 |
View Full Product Details