Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- 2810047C21Rik1
- Entrez Gene ID
-
72716
- Gene Name
- RIKEN cDNA 2810047C21 gene 1
- Gene Aliases
-
100042870, 2810047C21Rik, AA683836, Gm4082
- Chromosome Location
-
Chr.7: 9869968 - 9876963 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_015598.1 |
|
2 - 3 |
470 |
99 |
GenBank mRNA |
AK083498.1 |
- |
2 - 3 |
471 |
99 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BB654671.1 |
- |
2 - 3 |
468 |
|
Chr. 7 - 8088997 - 8093035 [-] on Build GRCm38 |
99 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
vomeronasal 2, receptor 47 |
Vmn2r47 |
|
-32.9kb
|
Sense |
Chr. - 9816782 - 9841325 on Build GRCm38 |
Downstream |
vomeronasal 2, receptor 48 |
Vmn2r48 |
EG625580 |
51.3kb
|
Sense |
Chr. - 9927624 - 9953585 on Build GRCm38 |
View Full Product Details