Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- 4921511I17Rik
- Entrez Gene ID
-
70858
- Gene Name
- RIKEN cDNA 4921511I17 gene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 12351522 - 12392475 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_045829.1 |
|
3 - 4 |
252 |
87 |
GenBank mRNA |
AK014875.1 |
- |
3 - 4 |
253 |
87 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BY714395.1 |
- |
3 - 4 |
253 |
|
Chr. 12 - 12351947 - 12392643 [-] on Build GRCm38 |
87 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
tubulin, beta 2a, pseudogene 2 |
Tubb2a-ps2 |
C79445 |
-471.1kb
|
Sense |
Chr. - 11882196 - 11882899 on Build GRCm38 |
Downstream |
predicted gene, 34637 |
Gm34637 |
|
294.6kb
|
Sense |
Chr. - 12667732 - 12669963 on Build GRCm38 |
View Full Product Details