Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfam1
Entrez Gene ID
74039
Gene Name
Nfat activating molecule with ITAM motif 1
Gene Aliases
4921501M20Rik, Cnaip
Chromosome Location
Chr.15: 82996736 - 83033397 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271411.1 NP_001258340.1 4 - 5 820 77
NM_001271412.1 NP_001258341.1 4 - 5 750 77
NM_001271413.1 NP_001258342.1 4 - 5 659 77
NM_001271414.1 NP_001258343.1 4 - 5 747 77
NM_028728.3 NP_083004.1 4 - 5 677 77
GenBank mRNA AF361364.1 - 4 - 5 754 77
AK014802.2 - 4 - 5 678 77
AK048992.1 - 4 - 5 661 77
AK150635.1 - 4 - 5 729 77
AK151187.1 - 4 - 5 729 77
AK151884.1 - 4 - 5 729 77
AK152262.1 - 4 - 5 729 77
AK152499.1 - 4 - 5 729 77
AK152688.1 - 4 - 5 729 77
AK155106.1 - 4 - 5 674 77
AK162939.1 - 4 - 5 731 77
AK169918.1 - 4 - 5 674 77
AK170098.1 - 4 - 5 764 77
AK170355.1 - 4 - 5 727 77
AK170545.1 - 4 - 5 615 77
AK170593.1 - 4 - 5 685 77
AK170677.1 - 4 - 5 685 77
AK170811.1 - 4 - 5 685 77
AK170883.1 - 4 - 5 728 77
AK171082.1 - 4 - 5 790 77
AK171086.1 - 4 - 5 698 77
AK171201.1 - 4 - 5 659 77

View Full Product Details