Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARMC1
Entrez Gene ID
101119911
Gene Name
armadillo repeat containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 46423965 - 46447313 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004011704.4 XP_004011753.2 3 - 4 505 76

View Full Product Details