Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BLVRB
Entrez Gene ID
101119750
Gene Name
biliverdin reductase B
Gene Aliases
-
Chromosome Location
Chr.14: 52898612 - 52916232 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978310.1 XP_027834111.1 1 - 2 216 80

View Full Product Details