Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCK
Entrez Gene ID
101116683
Gene Name
cholecystokinin
Gene Aliases
-
Chromosome Location
Chr.19: 15495178 - 15501079 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027958055.1 XP_027813856.1 1 - 2 1318 67

View Full Product Details