Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CA11
Entrez Gene ID
443426
Gene Name
carbonic anhydrase 11
Gene Aliases
-
Chromosome Location
Chr.14: 58736076 - 58741949 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009408.2 NP_001009408.1 1 - 2 308 104
GenBank mRNA Y07785.1 - - 309 104

View Full Product Details