Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRD1
Entrez Gene ID
101110226
Gene Name
leucine rich repeats and death domain containing 1
Gene Aliases
-
Chromosome Location
Chr.4: 11135748 - 11154916 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007706.4 XP_004007755.1 1 - 2 2531 143

View Full Product Details