Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHO2
Entrez Gene ID
101107604
Gene Name
pleckstrin homology domain containing O2
Gene Aliases
-
Chromosome Location
Chr.7: 46312366 - 46334771 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010530.4 XP_004010579.1 4 - 5 484 66

View Full Product Details