Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUMO3
Entrez Gene ID
101104960
Gene Name
small ubiquitin like modifier 3
Gene Aliases
-
Chromosome Location
Chr.1: 288021599 - 288031114 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027961105.1 XP_027816906.1 3 - 4 364 89

View Full Product Details