Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST8SIA3
Entrez Gene ID
101114742
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 3
Gene Aliases
-
Chromosome Location
Chr.23: 62701531 - 62719805 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015103786.2 XP_014959272.2 2 - 3 837 111

View Full Product Details