Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A15
Entrez Gene ID
101121050
Gene Name
membrane spanning 4-domains A15
Gene Aliases
-
Chromosome Location
Chr.21: 38159606 - 38168322 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015103121.2 XP_014958607.2 3 - 4 387 67
XM_027959729.1 XP_027815530.1 4 - 5 510 67

View Full Product Details