Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ICMT
Entrez Gene ID
101117687
Gene Name
isoprenylcysteine carboxyl methyltransferase
Gene Aliases
-
Chromosome Location
Chr.12: 50754907 - 50762039 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004014030.4 XP_004014079.3 2 - 3 330 87

View Full Product Details