Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAD51AP2
Entrez Gene ID
101106070
Gene Name
RAD51 associated protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 26907549 - 26914220 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004007214.4 XP_004007263.3 2 - 3 3345 129

View Full Product Details