Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DUSP8
Entrez Gene ID
101122905
Gene Name
dual specificity phosphatase 8
Gene Aliases
-
Chromosome Location
Chr.21: 51851715 - 51867567 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027959541.1 XP_027815342.1 2 - 3 411 87

View Full Product Details