Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MARCHF11
Entrez Gene ID
101102675
Gene Name
membrane associated ring-CH-type finger 11
Gene Aliases
MARCH11
Chromosome Location
Chr.16: 62170474 - 62286575 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012097221.2 XP_011952611.2 1 - 2 533 83

View Full Product Details