Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC101112936
Entrez Gene ID
101112936
Gene Name
interferon-activable protein 203
Gene Aliases
IFI16, IFI16l
Chromosome Location
Chr.1: 116579548 - 116622690 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004002644.4 XP_004002693.2 7 - 8 1529 124
GenBank mRNA KX931094.1 - 7 - 8 1531 124

View Full Product Details