Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC71
Entrez Gene ID
101105954
Gene Name
coiled-coil domain containing 71
Gene Aliases
-
Chromosome Location
Chr.19: 52842480 - 52847199 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957640.1 XP_027813441.1 1 - 2 242 95

View Full Product Details