Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDH12
Entrez Gene ID
101106077
Gene Name
protocadherin 12
Gene Aliases
-
Chromosome Location
Chr.5: 55350849 - 55364622 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015096111.2 XP_014951597.2 2 - 3 3295 94

View Full Product Details