Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAV3
Entrez Gene ID
101107228
Gene Name
caveolin 3
Gene Aliases
-
Chromosome Location
Chr.19: 18765985 - 18778596 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018300.4 XP_004018349.1 1 - 2 269 90

View Full Product Details