Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC101117458
Entrez Gene ID
101117458
Gene Name
UTP11 small subunit processome component
Gene Aliases
UTP11
Chromosome Location
Chr.1: 13243309 - 13254576 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012100981.3 XP_011956371.1 6 - 7 662 79

View Full Product Details