Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNIP3
Entrez Gene ID
101114957
Gene Name
TNFAIP3 interacting protein 3
Gene Aliases
-
Chromosome Location
Chr.6: 5192955 - 5251787 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027971373.1 XP_027827174.1 11 - 12 1055 67

View Full Product Details