Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GYG2
Entrez Gene ID
101115765
Gene Name
glycogenin 2
Gene Aliases
-
Chromosome Location
Chr.X: 1309073 - 1332765 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012106161.3 XP_011961551.2 3 - 4 766 96
XM_027962831.1 XP_027818632.1 3 - 4 761 96
XM_027962832.1 XP_027818633.1 5 - 6 776 96
XM_027962833.1 XP_027818634.1 5 - 6 768 96

View Full Product Details