Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IGSF8
Entrez Gene ID
101102283
Gene Name
immunoglobulin superfamily member 8
Gene Aliases
-
Chromosome Location
Chr.1: 117966985 - 117974018 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976987.1 XP_027832788.1 2 - 3 608 125

View Full Product Details