Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRC41
Entrez Gene ID
101112107
Gene Name
leucine rich repeat containing 41
Gene Aliases
-
Chromosome Location
Chr.1: 21529173 - 21547779 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015091953.2 XP_014947439.2 2 - 3 1218 130
XM_027969491.1 XP_027825292.1 2 - 3 299 130
XM_027969500.1 XP_027825301.1 2 - 3 2698 130

View Full Product Details