Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACO1
Entrez Gene ID
100009154
Gene Name
aconitase 1
Gene Aliases
FRP
Chromosome Location
Chr.1: 21028534 - 21086931 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082315.1 NP_001075784.1 17 - 18 2180 137
XM_008248341.2 XP_008246563.1 17 - 18 2242 137
XM_008253164.2 XP_008251386.1 16 - 17 2142 137
GenBank mRNA M95815.1 - 17 - 18 2180 137

View Full Product Details