Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS72
Entrez Gene ID
100328667
Gene Name
vacuolar protein sorting 72 homolog
Gene Aliases
-
Chromosome Location
Chr.13: 41009573 - 41016769 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171300.1 NP_001164771.1 3 - 4 559 86

View Full Product Details