Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HBG1
Entrez Gene ID
100328859
Gene Name
hemoglobin, gamma A
Gene Aliases
HBE1
Chromosome Location
Chr.1: 146261868 - 146263244 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171504.1 NP_001164975.1 2 - 3 311 73

View Full Product Details