Gene Transcripts

Gene Symbol
Dnah8
Entrez Gene ID
207117
Gene Name
dynein, axonemal, heavy chain 8
Gene Aliases
Dlp8, Dnahc8
Chromosome Location
Chr. 20: 9301314 - 9560805 on Build Rnor_6.0
UniGene ID
Rn.224155
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008772852.1 XP_008771074.1 83 12829

View Full Product Details