Gene Transcripts

Gene Symbol
Ms4a4c
Entrez Gene ID
293746
Gene Name
membrane-spanning 4-domains, subfamily A, member 4C
Gene Aliases
Chromosome Location
Chr. 1: 227640733 - 227661731 on Build Rnor_6.0
UniGene ID
Rn.218548
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106339.1 NP_001099809.1 1 58
XM_006231087.2 XP_006231149.1 2 227
XM_006231088.2 XP_006231150.1 2 227

View Full Product Details