Gene Transcripts

Gene Symbol
Atp2b1
Entrez Gene ID
29598
Gene Name
ATPase, Ca++ transporting, plasma membrane 1
Gene Aliases
Pmca1a, Pmca1b, Pmca1c
Chromosome Location
Chr. 7: 41114606 - 41223116 on Build Rnor_6.0
UniGene ID
Rn.228999
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053311.1 NP_445763.1 11 1945
GenBank mRNA J03753.1 - 11 1945

View Full Product Details