Gene Transcripts

Gene Symbol
Dsel
Entrez Gene ID
297865
Gene Name
dermatan sulfate epimerase-like
Gene Aliases
RGD1307724
Chromosome Location
Chr. 13: 1942384 - 1948079 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006249608.2 XP_006249670.1 1 3505

View Full Product Details