Gene Transcripts

Gene Symbol
Actr5
Entrez Gene ID
362258
Gene Name
ARP5 actin-related protein 5 homolog (yeast)
Gene Aliases
Chromosome Location
Chr. 3: 155118567 - 155132073 on Build Rnor_6.0
UniGene ID
Rn.51809
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108600.1 NP_001102070.1 4 1029
XM_006235422.2 XP_006235484.1 4 1050

View Full Product Details