Gene Transcripts

Gene Symbol
Mesdc2
Entrez Gene ID
308796
Gene Name
mesoderm development candidate 2
Gene Aliases
Chromosome Location
Chr. 1: 146037426 - 146043097 on Build Rnor_6.0
UniGene ID
Rn.41045
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008345.1 NP_001008346.1 3 725
GenBank mRNA BC085892.1 - 3 725

View Full Product Details