Gene Transcripts

Gene Symbol
Lyc2
Entrez Gene ID
688047
Gene Name
lysozyme C type 2
Gene Aliases
Lyz2
Chromosome Location
Chr. 7: 60316045 - 60323610 on Build Rnor_6.0
UniGene ID
Rn.162941
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128494.1 NP_001121966.1 1 313
XM_006241392.2 XP_006241454.1 1 137
GenBank mRNA BC166737.1 - 1 313

View Full Product Details