Gene Transcripts

Gene Symbol
Sh3bgrl3
Entrez Gene ID
298544
Gene Name
SH3 domain binding glutamate-rich protein like 3
Gene Aliases
Chromosome Location
Chr. 5: 152357270 - 152358643 on Build Rnor_6.0
UniGene ID
Rn.18564
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106688.1 NP_001100158.1 2 222
GenBank mRNA BC167002.1 - 2 147

View Full Product Details