Gene Transcripts

Gene Symbol
Tlcd2
Entrez Gene ID
497955
Gene Name
TLC domain containing 2
Gene Aliases
RGD1562631
Chromosome Location
Chr. 10: 62303124 - 62306198 on Build Rnor_6.0
UniGene ID
Rn.163153
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006246868.2 XP_006246930.1 4 1275
XM_573142.6 XP_573142.4 4 1275
GenBank mRNA FQ219135.1 - 4 747

View Full Product Details