Gene Transcripts

Gene Symbol
Itpripl2
Entrez Gene ID
499253
Gene Name
inositol 1,4,5-trisphosphate receptor interacting protein-like 2
Gene Aliases
RGD1564255
Chromosome Location
Chr. 1: 188140853 - 188142748 on Build Rnor_6.0
UniGene ID
Rn.140535
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127303.1 NP_001120775.1 1 181
GenBank mRNA BC166746.1 - 1 181

View Full Product Details