Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APOBEC2
Entrez Gene ID
719677
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 2
Gene Aliases
EGK_14876
Chromosome Location
Chr.4: 128340926 - 128352664 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015136306.2 XP_014991792.1 - 279 121

View Full Product Details