Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FCRL2
Entrez Gene ID
718824
Gene Name
Fc receptor like 2
Gene Aliases
-
Chromosome Location
Chr.1: 93212631 - 93243025 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001116902.4 XP_001116902.3 - 2752 66

View Full Product Details