Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AIG1
Entrez Gene ID
700988
Gene Name
androgen induced 1
Gene Aliases
-
Chromosome Location
Chr.4: 46841323 - 47116713 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260565.1 NP_001247494.1 - 189 70
XM_028846705.1 XP_028702538.1 - 387 70
XM_028846706.1 XP_028702539.1 - 223 70
XM_028846707.1 XP_028702540.1 - 303 70
XM_028846708.1 XP_028702541.1 - 229 70
GenBank mRNA JU328131.1 - - 120 70
JV047297.1 - - 189 70
JV601698.1 - - 182 70

View Full Product Details