Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ADGRE2
Entrez Gene ID
654420
Gene Name
adhesion G protein-coupled receptor E2
Gene Aliases
EMR2
Chromosome Location
Chr.19: 14368627 - 14412439 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038662.1 NP_001033751.1 - 42 63
XM_028838330.1 XP_028694163.1 - 196 63
XM_028838331.1 XP_028694164.1 - 196 63
XM_028838333.1 XP_028694166.1 - 199 63
GenBank mRNA DQ227271.1 - - 42 63

Gene Symbol
ADGRE5
Entrez Gene ID
100426363
Gene Name
adhesion G protein-coupled receptor E5
Gene Aliases
CD97
Chromosome Location
Chr.19: 13993332 - 14021236 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015123131.2 XP_014978617.1 - 400 63
XM_015123132.2 XP_014978618.1 - 400 63
XM_015123133.2 XP_014978619.1 - 399 63
XM_015123134.2 XP_014978620.1 - 400 63
XM_028839871.1 XP_028695704.1 - 400 63
XM_028839872.1 XP_028695705.1 - 400 63
XM_028839873.1 XP_028695706.1 - 234 63
GenBank mRNA JU478010.1 - - 117 63
JU478011.1 - - 117 63

View Full Product Details