Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SVBP
- Entrez Gene ID
-
698308
- Gene Name
- small vasohibin binding protein
- Gene Aliases
-
CCDC23, EGK_00646
- Chromosome Location
-
Chr.1: 181738104 - 181751240 on Build Mmul_10
- Gene Symbol
- ERMAP
- Entrez Gene ID
-
698433
- Gene Name
- erythroblast membrane associated protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 181703294 - 181723798 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JU332797.1 |
- |
- |
411 |
67 |
JV706212.1 |
- |
- |
340 |
67 |
View Full Product Details