Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NMRK1
Entrez Gene ID
703868
Gene Name
nicotinamide riboside kinase 1
Gene Aliases
C15H9orf95
Chromosome Location
Chr.15: 90493972 - 90522406 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261283.1 NP_001248212.1 - 758 132
XM_015117794.2 XP_014973280.1 - 849 132
XM_028834280.1 XP_028690113.1 - 584 132
GenBank mRNA JU323108.1 - - 830 132
JU323213.1 - - 758 132
JV717796.1 - - 225 132

View Full Product Details