Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC71
Entrez Gene ID
707774
Gene Name
coiled-coil domain containing 71
Gene Aliases
EGK_11645
Chromosome Location
Chr.2: 105242156 - 105245861 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266198.1 NP_001253127.1 - 72 88
GenBank mRNA JU476847.1 - - 72 88
JV659107.1 - - 79 88
JV721385.1 - - 82 88

View Full Product Details