Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TKTL2
Entrez Gene ID
706986
Gene Name
transketolase like 2
Gene Aliases
-
Chromosome Location
Chr.5: 161089908 - 161092706 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001095403.4 XP_001095403.2 - 205 113

View Full Product Details