Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRC1
Entrez Gene ID
701010
Gene Name
mannose receptor C-type 1
Gene Aliases
MRC1L1
Chromosome Location
Chr.9: 18959139 - 19056976 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193925.3 NP_001180854.2 - 2739 108

View Full Product Details